missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Humidity absorber, Solstice

Product Code. 15634410
Change view
Click to view available options
Quantity:
500 g
1 kg
6 x 500 g
6 x 1 kg
Unit Size:
1kg
500g
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15634410 500 g 500g
15624410 1 kg 1kg
15654410 6 x 500 g Pack of 6
15644410 6 x 1 kg Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15634410 Supplier Honeywell-Hydranal Supplier No. 34788500G
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.
View alternative products
This item is not returnable. View return policy

Drying agent for air and gases for Karl Fischer titration (with indicator)

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Humidity absorber
CAS 7631-86-9
Quantity 500 g
UN Number NONH for all modes of transport
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.