missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Imidazole, Solstice

Product Code. 15655170
Change view
Click to view available options
Quantity:
500 g
Unit Size:
500g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15655170 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15655170 Supplier Honeywell-Hydranal Supplier No. 37864500G
  • ONLINE EXCLUSIVE PRICE
    126.00 €
    116.81 € / 500g
    Save 9.19 €
    7% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Buffer substance for Karl Fischer titration

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Imidazole
Packaging Plastic Bottle
CAS 288-32-4
Quantity 500 g
Flash Point 145°C (293°F)
UN Number UN3263
compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL Imidazole
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Reproductive toxicity Category 1B
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H360D-May damage the unborn child.
Precautionary Statement
  • P201-Obtain special instructions before use.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: imidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.