missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - KetoSolver, Reagent for volumetric one-component KF titration in aldehydes and ketones (working medium), Honeywell Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
6 x 500 mL
6 x 1 L
Unit Size:
500mL
Pack of 6
Description
Advantages of Hydranal one-component reagents:
- Unlimited water capacity
- Convenient and easy to use
- The greatest flexibility in working media selection
- reduced toxicity compared to methanol based systems
- Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
- Long shelf life (three years)
Improved Composition
Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.
Specifications
Specifications
| Chemical Name or Material | HYDRANAL™-KetoSolver |
| CAS | 288-32-4, 64-17-5, 107-98-2, 101023-55-6 |
| Density | 0.91g/mL |
| Quantity | 6 x 500 mL |
| Flash Point | 21°C (69.8°F) |
| UN Number | UN1993BE |
Safety and Handling
Product Identifier
- HYDRANAL-KetoSolver
- Danger
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 3
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H336-May cause drowsiness or dizziness.
- H360D-May damage the unborn child.
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : 1-methoxy-2-propanol; monopropylene glycol methyl ether, ethanol; ethyl alcohol,imidazole, Imidazol
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction