missing translation for 'onlineSavingsMsg'
Learn More
Learn More
HYDRANAL™ - Methanol dry, Reagent for volumetric one-component KF titration (working medium), Honeywell Fluka™
26.55 € - 183.51 €
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15644390
|
Honeywell-Hydranal
34741-1L |
1 L |
26.55 €
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15654390
|
Honeywell-Hydranal
34741-2.5L |
2.5 L |
52.56 €
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15674390
|
Honeywell Fluka
34741-6X1L |
6 x 1 L |
137.31 €
Pack of 6 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
15664390
|
Honeywell Fluka
34741-4X2.5L |
4 x 2.5 L |
183.51 €
Pack of 4 |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Safety and Handling
P210-P280-P302 + P352 + P312-P304 + P340 + P312-P370 + P378-P403 + P235
missing translation for 'rtecsNumber' : PC1400000
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- HYDRANAL-Methanol dry
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Acute toxicity Category 3
- Specific target organ toxicity after single exposure Category 1
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H301-Toxic if swallowed.
- H311-Toxic in contact with skin.
- H331-Toxic if inhaled.
- H370-Causes damage to organs.
Precautionary Statement
- P260-Do not breathe dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P284-¢In case of inadequate ventilation! wear respiratory protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
- MIXTURE LIST-Contains : methanol