missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - NEXTGEN Coulomat A-FA, Solstice

Product Code. 17504956
Change view
Click to view available options
Quantity:
1 x 500 mL
6 x 500 mL
Unit Size:
500mL
6 bottles
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17504956 6 x 500 mL 6 bottles
17893080 1 x 500 mL 500mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 17504956 Supplier Hydranal Supplier No. 344716X500ML
  • ONLINE EXCLUSIVE PRICE
    2575.00 €
    2403.49 € / Pack of 6
    Save 171.51 €
    6% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Anolyte for coulometric Karl Fischer titration of ketones and Li-ion battery electrolytes, preferred for cells with diaphragm, acetonitrile-based, free of alcohols (free of CMR substances)

  • HYDRANAL™ is the most trusted brand in Karl Fischer titration because of its dedication to quality, consistency and continued innovation.
    • Comprehensive product portfolio with 60 available products
    • 40 years of innovation – check our NEXTGEN reagents HERE
    • Dedicated technical service and support
    • More than 800 Application Notes available on our website
    • View our NEXTGEN Coulomat A-FA standard operating procedure
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™ - NEXTGEN Coulomat A-FA
CAS Min % ≥2.00, ≥60.00
CAS Max % <70.00, <10.00, <10.00
Color Orange
Density 0.906 g/cm3
CAS Confidential Stabilizer 2, 75-75-2, 75-05-8
pH 5.5
Physical Form Liquid
Quantity 6 x 500 mL
Flash Point 12°C to 13°C
Packaging Glass Bottle
UN Number UN2924
Solubility Information Soluble in water
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL NEXTGEN Coulomat A-FA
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H290-May be corrosive to metals.
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H332-Harmful if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: acetonitrile, methanesulphonic acid, 1-Ethylimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.