missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - NEXTGEN Solvent E-FI, Solstice

Product Code. 16892360
Change view
Click to view available options
Quantity:
1 L
6 x 1 L
Unit Size:
1 L
6 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16892360 6 x 1 L 6 L
16882360 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16892360 Supplier Hydranal Supplier No. 344316X1L
  • ONLINE EXCLUSIVE PRICE
    735.00 €
    684.97 € / 6 L
    Save 50.03 €
    6% Off
 Restricted Item
Estimated Shipment: 31-03-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Medium for volumetric, two-component Karl Fischer titrations

  • Ethanol-based
  • Free of Imidazole
  • Free of CMR Substances
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™ - NEXTGEN Solvent E-FI
Melting Point -115°C
CAS Min % ≥55%
Color Colorless
Density 0.895 g/cm3
pH 5 to 6
Physical Form Liquid
CAS 64-17-5
Quantity 6 x 1 L
CAS Max % ≤80%
Packaging Glass Bottle
UN Number UN1170
Solubility Information Completely Miscible in Water
Grade Karl Fischer
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • HYDRANAL NEXTGEN Solvent E-FI
Signal Word
  • Danger
Hazard Category
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: ethanol, 2-ethykimidazole, sulphur dioxide, L-proline, 2-ethyl imidazole hydroiodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.