missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Sodium tartrate dihydrate, Solstice

Product Code. 15624430
Change view
Click to view available options
Quantity:
25 g
100 g
6 x 100 g
Unit Size:
100g
25g
Pack of 6
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15624430 100 g 100g
15644430 25 g 25g
15664430 6 x 100 g Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15624430 Supplier Honeywell-Hydranal Supplier No. 34803100G
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Test substance for volumetric Karl Fischer titration (water content ∼15.66%, exact value on report of analysis)

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Sodium tartrate dihydrate
CAS 6106-24-7
Quantity 100 g
UN Number NONH for all modes of transport
Grade Karl Fischer
Product Identifier
  • HYDRANAL Sodium tartrate dihydrate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.