missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Solvent E, Honeywell Fluka™

Product Code. 15624370 Shop All Honeywell Fluka Products
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
500mL
Pack of 4
Pack of 6
This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Product Code. 15624370

Brand: Honeywell-Hydranal 34730500ML

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains a substance listed in Annex XIV to REACH Regulation (EC) No 1907/2006, all customers must complete a declaration before delivery.

Medium for volumetric two-component Karl Fischer titration (ethanol-based)

Advantages of Hydranal two-component reagents:

  • High titration speed
  • Ideal accuracy for small amounts of water
  • High buffer capacity
  • Exact and stable titer
  • E-type reagents: reduced toxicity compared to methanol
  • Long shelf life (three years for titrants, five years for solvents)

Improved Composition

In two-component reagents the Karl Fisher reactants are separated into two solutions: the titrant and the solvent. Hydranal-Titrant contains iodine dissolved in an alcohol with a precisely defined concentration. Hydranal-Solvent is an alcoholic solution of sulfur dioxide and imidazole. The alcohol is either methanol for standard reagents or ethanol for E-type reagents.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Solvent E
CAS 288-32-4, 111-42-2, 64-17-5, 7446-09-5
Density 0.90g/mL
Quantity 500 mL
Flash Point 12°C (53.6°F)
MDL Number MFCD01867183
UN Number UN1170
compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL Solvent E
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Serious eye damage/eye irritation Category 1
  • Reproductive toxicity Category 1B
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H318-Causes serious eye damage.
  • H360D-May damage the unborn child.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-¢In case of inadequate ventilation! wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : ethanol; ethyl alcohol, 2,2'-iminodiethanol; diethanolamine,imidazole, sulphurdioxide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.