missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Solver (Crude) oil, Solstice

Product Code. 15694350
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
Pack of 4
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15694350 6 x 1 L Pack of 6
15664350 1 L 1 L
15674350 2.5 L 2.5 L
15684350 4 x 2.5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15694350 Supplier Honeywell-Hydranal Supplier No. 346976X1L
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Medium for volumetric one- and two-component Karl Fischer titration in oils (chloroform-methanol-xylene-based)

Advantages of Hydranal one-component reagents:

  • Unlimited water capacity
  • Convenient and easy to use
  • The greatest flexibility in working media selection
  • reduced toxicity compared to methanol based systems
  • Suitable for methanol-reacting compounds, e.g. ketones and aldehydes
  • Long shelf life (three years)

Improved Composition

Hydranal-Composite contains all the reactants including iodine, sulfur dioxide, and the bases imidazole and 2-methylimidazole, dissolved in diethylene glycol monoethyl ether (DEGEE). Adding 2-methylimidazole in addition to imidazole improves the stability and eliminates the formation of crystals which can interfere with the titrator’s performance.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Solver (Crude) oil
CAS 67-56-1, 7446-09-5, 67-66-3, 95-47-6, 288-32-4
Density 1.01g/mL
Quantity 6 x 1 L
Flash Point 6°C (42.8°F)
UN Number UN1992
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.