missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Standard 5.0, Solstice

Product Code. 15634470
Change view
Click to view available options
Quantity:
100 mL
500 mL
6 x 100 mL
6 x 500 mL
Unit Size:
100mL
500mL
Pack of 6
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15634470 100 mL 100mL
15644470 500 mL 500mL
15654470 6 x 100 mL Pack of 6
15664470 6 x 500 mL Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 15634470 Supplier Honeywell-Hydranal Supplier No. 34813100ML
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Test solution for rough check of volumetric KF equipment.

Advantages of HYDRANAL- Auxiliaries

  • Broad product range of solubilizer for special sample application
  • Broad product range of buffer for pH related sample application
  • Drying agents for cell protection and drying of carrier gasses.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Standard 5.0
Packaging Glass Bottle
CAS 71-36-3, 95-47-6
Quantity 100 mL
Flash Point 27°C (80.6°F)
UN Number UN1993C
Grade Karl Fischer
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL-Standard 5.0
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Aspiration toxicity Category 1
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H412-Harmful to aquatic life with long lasting effects.
  • H302-Harmful if swallowed.
  • H304-May be fatal if swallowed and enters airways.
  • H312-Harmful in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H332-Harmful if inhaled.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: o-xylene, butan-1-ol; n-butanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.