missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Titrant 5 E, Honeywell Fluka™

Product Code. 15684370 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
4 x 2.5 L
Unit Size:
Pack of 4
Product Code. Quantity unitSize
15684370 4 x 2.5 L Pack of 4
1 options
This item is not returnable. View return policy

Product Code. 15684370

Brand: Honeywell Fluka 347324X2.5L

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Titrating agent for volumetric two-component Karl Fischer titration (ethanol-based)

Hydranal™ is the most trusted brand in Karl Fischer titration because of its dedication to quality, consistency and continued innovation.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™ - Titrant 5 E
CAS 7553-56-2, 68007-08-9, 64-17-5
Quantity 4 x 2.5 L
UN Number UN1170
compliance-icons compliance-icons
Product Identifier
  • HYDRANAL Titrant 5 E
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : ethanol, iodine, 1H-imidazole monohydriodide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.