missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Titrant 5, Reagent for volumetric two-component Karl Fischer titration, Solstice

Product Code. 15664420
Change view
Click to view available options
Quantity:
500 mL
1 L
2.5 L
6 x 500 mL
4 x 2.5 L
6 x 1 L
Unit Size:
1 L
2.5 L
500mL
Pack of 4
Pack of 6
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15664420 500 mL 500mL
15634420 1 L 1 L
15644420 2.5 L 2.5 L
15684420 6 x 500 mL Pack of 6
15674420 6 x 1 L Pack of 6
15654420 4 x 2.5 L Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Product Code. 15664420 Supplier Honeywell-Hydranal Supplier No. 34801500ML
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 67-56-1

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Titrant 5
Density 0.85g/mL
Packaging Glass Bottle
Quantity 500 mL
Flash Point 11°C (51.8°F)
UN Number UN1230
Grade Karl Fischer
compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL Titrant 5
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Acute toxicity Category 3
  • Specific target organ toxicity after single exposure Category 1
  • Specific target organ toxicity after repeated exposure Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301+H311+H331-Toxic if swallowed, in contact with skin or if inhaled
  • H370-Causes damage to organs.
  • H373-May cause damage to organs.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: methanol, iodine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.