missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Water Standard 0.1, Standard for Karl Fischer titration (water content 0.1 mg/g = 0.01%), verified against NIST SRM 2890 & NMIJ CRM 4222, Solstice

Product Code. 15624510
Change view
Click to view available options
Quantity:
1 Kit
10 x 4 mL, Ampules
Unit Size:
40mL
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15624510 1 Kit Pack of 6
15648380 10 x 4 mL, Ampules 40mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.
Product Code. 15624510 Supplier Honeywell-Hydranal Supplier No. 348476X40ML
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Regulatory: This product is subject to excise duty on mineral oils in accordance with European Directive 2003/96/EC.

We regret to inform you that this product is not available for the Canary Islands, Ceuta and Melilla. We apologise for the inconvenience.

C8H10 | 106.17 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008519
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
IUPAC Name 1,2-xylene
SMILES CC1=CC=CC=C1C

Specifications

Boiling Point 136°C to 145°C
Density 0.86 g/mL at 20 °C
CAS 7732-18-5, 95-47-6
Quantity 1 Kit
CAS Max % ≤100.0000%
Flash Point 23 − 27°C
Packaging Ampoule
UN Number UN1307
Formula Weight 106.17
Grade Karl Fischer
Chemical Name or Material HYDRANAL™ - Water Standard 0.1, Standard for Karl Fischer titration (water content 0.1 mg/g = 0.01%), exact value on report of analysis
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL Water Standard 0.1
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Acute toxicity Category 4
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Aspiration toxicity Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H312+H332-Harmful in contact with skin or if inhaled
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: o-xylene
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.