missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Water Standard 1.0, Standard for Karl Fischer titration (water content 1 mg/g = 0.1%), verified against NIST SRM 2890 & NMIJ CRM 4222, Solstice

Product Code. 15628380
Change view
Click to view available options
Quantity:
1 Kit
40 mL
Unit Size:
40mL
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15628380 40 mL 40mL
15664490 1 Kit Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15628380 Supplier Honeywell-Hydranal Supplier No. 3482840ML
  • ONLINE EXCLUSIVE PRICE
    94.00 €
    86.90 € / 40mL
    Save 7.10 €
    7% Off
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 100-66-3, 108-32-7, 108-32-7, 100-66-3
Quantity 40 mL
Flash Point 43°C (109.4°F)
UN Number UN2222
Grade Certified
Chemical Name or Material HYDRANAL™-Water Standard 1.0
compliance-icons
Product Identifier
  • HYDRANAL
Signal Word
  • Danger||Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Anisole, propylene carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.