missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Water Standard 1.0, Standard for Karl Fischer titration (water content 1 mg/g = 0.1%), verified against NIST SRM 2890 & NMIJ CRM 4222, Solstice

Product Code. 15664490
Change view
Click to view available options
Quantity:
1 Kit
40 mL
Unit Size:
40mL
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15664490 1 Kit Pack of 6
15628380 40 mL 40mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15664490 Supplier Honeywell-Hydranal Supplier No. 348286X40ML
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

Molecular Formula C7H8O
Molecular Weight (g/mol) 108.14
MDL Number MFCD00007941
InChI Key RDOXTESZEPMUJZ-UHFFFAOYSA-N
Synonym Methoxybenzene, Methyl phenyl ether
PubChem CID 7519
ChEBI CHEBI:16579
IUPAC Name anisole
SMILES COC1=CC=CC=C1

Specifications

Boiling Point 153°C to 155°C
CAS Min % ≥90.0000%, ≥1.0000%
Density 1.000 g/cm3
CAS 108-32-7, 100-66-3
Quantity 1 Kit
CAS Max % <10.0000%
Flash Point 43°C
Linear Formula HCONH2
Packaging Ampoule
UN Number UN2222
Grade Karl Fischer
Chemical Name or Material HYDRANAL™ - Water Standard 1.0, Standard for Karl Fischer titration (water content 1 mg/g = 0.1%), exact value on report of analysis
Show More Show Less
compliance-icons
Product Identifier
  • HYDRANAL Water Standard 1.3
Signal Word
  • Warning
Hazard Category
  • Flammable liquid Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
Supplemental information
  • MIXTURE LIST-Contain: Anisole, propylene carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.