missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Water Standard 10.0, Standard for Karl Fischer titration (water content 10 mg/g = 1.0%), Solstice

Product Code. 15634510
Change view
Click to view available options
Quantity:
10 x 8 mL
6 x (10 x 8 mL)
Unit Size:
80mL
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15634510 6 x (10 x 8 mL) Pack of 6
15644510 10 x 8 mL 80mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below
Product Code. 15634510 Supplier Honeywell-Hydranal Supplier No. 348496X80ML
Discount Lab Supplies
Use promo code 28170
View all promotions
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
You may only order up to the available stock. Where an alternative item has been identified it will be shown below

CAS: 108-32-7 | C8H10 | 106.17 g/mol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

Molecular Formula C8H10
Molecular Weight (g/mol) 106.17
MDL Number MFCD00008519
InChI Key CTQNGGLPUBDAKN-UHFFFAOYSA-N
Synonym 1,2-Propanediol cyclic carbonate, 4-Methyl-1,3-dioxolan-2-one
PubChem CID 962
ChEBI CHEBI:15377
IUPAC Name 1,2-xylene
SMILES CC1=CC=CC=C1C

Specifications

CAS 108-32-7, 71-36-3, 95-47-6, 108-32-7
Boiling Point 109°C
CAS Min % ≥20.0000%
CAS Max % <30.0000%
Density 1.00 g/mL at 20 °C
Quantity 6 x (10 x 8 mL)
Flash Point 32°C
Packaging Ampoule
UN Number UN1993
Grade Karl Fischer
Assay effective water
Chemical Name or Material HYDRANAL™ - Water Standard 10.0, Standard for Karl Fischer titration (water content 10 mg/g = 1.0%), exact value on report of analysis
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL Water Standard 10.0
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: propylene carbonate,o-xylene, butan-1-ol; n-butanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.