missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™ - Water Standard 10.0, Standard for Karl Fischer titration (water content 10 mg/g = 1.0%), Solstice

Product Code. p-7122074
Change view
Click to view available options
Quantity:
10 x 8 mL
6 x (10 x 8 mL)
Unit Size:
80mL
Pack of 6
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15644510 10 x 8 mL 80mL
15634510 6 x (10 x 8 mL) Pack of 6
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15644510 Supplier Honeywell-Hydranal Supplier No. 3484980ML
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 111-46-6, 71-36-3, 95-47-6, 108-32-7
Density 1.00g/mL at 20°C
Quantity 10 x 8 mL
Flash Point 32°C (89.6°F)
Packaging Container
UN Number UN1993C
Grade Karl Fischer
Chemical Name or Material HYDRANAL™-Water Standard 10.0
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • HYDRANAL
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H336-May cause drowsiness or dizziness.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P284-[In case of inadequate ventilation] wear respiratory protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: propylene carbonate,o-xylene, butan-1-ol; n-butanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.