missing translation for 'onlineSavingsMsg'
Learn More

HYDRANAL™-Water Standard Oil, Solstice

Product Code. 15634350
Change view
Click to view available options
Quantity:
80 mL
Unit Size:
80mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15634350 80 mL 80mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15634350 Supplier Honeywell-Hydranal Supplier No. 3469480ML
ChemsChrom Mega Deals
Use promo code 27993
View all promotions
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Oil-based standard specifically designed for accurate Karl Fischer titration with a water content of less than 50 ppm.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material HYDRANAL™-Water Standard Oil
CAS 8042-47-5
Density 0.85g/mL at 20°C
Packaging Glass Bottle
Quantity 80 mL
Flash Point >112°C (>233.6°F)
UN Number NONH for all modes of transport
Product Identifier
  • HYDRANAL Water Standard Oil
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.