missing translation for 'onlineSavingsMsg'
Learn More

Hydrazine hydrate solution, 78-82% (iodometric), Honeywell Fluka™

Product Code. 15681650 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
250 mL
1 L
Unit Size:
1 L
250mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15681650 250 mL 250mL
15671650 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15681650 Supplier Honeywell-Fluka Supplier No. 18412250ML
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 10217-52-4
Molecular Formula H4N2
Molecular Weight (g/mol) 32.05
MDL Number MFCD00149931
InChI Key OAKJQQAXSVQMHS-UHFFFAOYSA-N
Synonym hydrazine hydrate, hydrazine monohydrate, hydrazinium hydroxide, hydrazine hydroxide, hydrazine, monohydrate, hydrazine, hydrate, idrazina idrata italian, hydrazinemonohydrate, unii-kyd297831p, ccris 7739
PubChem CID 24654
ChEBI CHEBI:35511
IUPAC Name hydrazine
SMILES NN

Specifications

Assay Percent Range 78 to 82%
Linear Formula NH2NH2 · H2O
UN Number UN2030
Beilstein 878137
Formula Weight 50.06
Percent Purity 78 to 82% (iodometric)
Grade Organics
Packaging Plastic Bottle
Quantity 250 mL
Assay acidimetric
Chemical Name or Material Hydrazine hydrate solution, 78-82% acidimetric
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.