missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Alfa Aesar™ Hydrindantin dihydrate, 98%
missing translation for 'orderingAttributeHoverText'
Quantity:
25 g
100 g
missing translation for 'unitSize'
100g
25g
Descrizione
Applications
Hydrindantin dihydrate is a reagent for photometric determination of amino acids and also used in amino acid determinations as a ninhydrin synergist. It is a chemical used in Stein-Moore-Chromatography.
Solubility
Insoluble in water. Soluble with decomposition in aqueous sodium carbonate (deep red color) and in sodium hydroxide solutions (deep blue color).
Notes
Store under cool dry place. Ensure good ventilation. Incompatible with oxidizing agents.
Hydrindantin dihydrate is a reagent for photometric determination of amino acids and also used in amino acid determinations as a ninhydrin synergist. It is a chemical used in Stein-Moore-Chromatography.
Solubility
Insoluble in water. Soluble with decomposition in aqueous sodium carbonate (deep red color) and in sodium hydroxide solutions (deep blue color).
Notes
Store under cool dry place. Ensure good ventilation. Incompatible with oxidizing agents.
Specifica
Specifica
| Melting Point | ∼250°C (decomposition) |
| Quantity | 25 g |
| Beilstein | 1895666 |
| Merck Index | 14,4775 |
| Formula Weight | 358.30 (322.27 Anhydrous) |
| Percent Purity | 98% |
| Chemical Name or Material | Hydrindantin dihydrate |
Correzione del contenuto del prodotto
Fornite il vostro feedback sul contenuto del prodotto compilando il modulo sottostante.
Titolo del prodotto
Individuate un'opportunità di miglioramento?Condividi una correzione di contenuto