missing translation for 'onlineSavingsMsg'
Learn More

Hydrochloric Acid Calibration and Matrix Blank For ICP/MS, SPEX CertiPrep™

Product Code. 15698265 Shop All SPEX Certiprep Products
Change view
Click to view available options
Quantity:
125 mL
Unit Size:
125mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15698265 125 mL 125mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15698265 Supplier SPEX Certiprep Supplier No. CLBLKHCL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Hydrochloric Acid Matrix Blank for ICP-MS, 125 mL in 2%HCl, SPEX CertiPrep™. ISO Guide 34 Certified, NIST Traceable Certified Reference Materials. Supplied with comprehensive Certificate of Analysis.

Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation

  • Hydrochlorid Acid Matrix
  • 125 mL volume

Certifications

ISO/IEC 17025:2005, ISO 17034:2016, ISO 9001: 2015

Compliance

GHS/OSHA Complaint

Specifications

Packaging Triple-leached LDPE bottle
CAS 7647-01-0
Certificate of Analysis Claritas PPT Certificate of Analysis reports up to 68 elements at ppt levels.
Concentration or Composition Notes Up to 68 elements are scanned with found values for ICP and ICP-MS standards. The found value of the trace metallic impurities is reported on the Certificate of Analysis.
Molecular Formula ClH
MDL Number MFCD00011324 MFCD00792839
For Use With (Equipment) AA, ICP, ICP/MS, IC, XRF, and Other Analytical Instrumentation
Synonym Muriatic Acid
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Type Certified Reference Material
SMILES Cl
IUPAC Name hydrogen chloride
Solvent or Matrix 2% HCl
Molecular Weight (g/mol) 36.46
PubChem CID 313
Quantity 125 mL
ChEBI CHEBI:17883
Grade ICP/ICP-MS
Chemical Name or Material Hydrochloric Acid Blank, 2% HCl in ASTM Type 1 Water
Show More Show Less

Product intended for laboratory use only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.