missing translation for 'onlineSavingsMsg'
Learn More

Hydrocoumarin, TRC

Product Code. 30392350
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30392350 25 g
30398921 5 g
30400492 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30392350 Supplier Toronto Research Chemicals Supplier No. TRCH71455525G
 Restricted Item
Estimated Shipment: 21-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-84-6
Molecular Formula C9 H8 O2
Molecular Weight (g/mol) 148.16
Synonym Hydrocoumarin (6CI,7CI,8CI), 3,4-Dihydro-2H-1-benzopyran-2-one, Hydrocinnamic acid, o-hydroxy-, δ-lactone (6CI), 2-Chromanone, 3,4-Dihydro-1H-benzopyran-2-one, 3,4-Dihydrocoumarin, D 1223, DHC, Dihydrocoumarin, Melilotin, Melilotin (coumarin), Melilotol
IUPAC Name 3,4-dihydrochromen-2-one
SMILES O=C1CCc2ccccc2O1

Specifications

Quantity 25 g
InChI Formula InChI=1S/C9H8O2/c10-9-6-5-7-3-1-2-4-8(7)11-9/h1-4H,5-6H2
Formula Weight 148.0524
Chemical Name or Material 3,4-Dihydrocoumarin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.