missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen chloride, 1M in diethyl ether

Product Code. 11496220
Change view
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
Product Code. Quantity unitSize
11496220 100 mL 100mL
11406230 500 mL 500mL
2 options
This item is not returnable. View return policy

Product Code. 11496220

Brand: Thermo Scientific Alfa Aesar H27125.AE

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7647-01-0 | ClH | 36.46 g/mol

In chemical industry production of inorganic salts, Production of vinyl chloride from acetylene, alkyl chlorides from olefins, dyes, artificial silk, pigment, s, pharmaceutical hydrochloride, As catalyst and solvent in organic syntheses, Alcohol chlorination reagent, Nitration reaction, Manufacture of phosphoric acid and in the production of ammonium chloride, Neutralization of basic systems, Cleaning agents. Hydrolyzing of starch and proteins in the preparation of various food products. The hydrogen chloride gas is mainly used for EDC and vinyl chloride monomer (VCM) production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7647-01-0
Molecular Formula ClH
Molecular Weight (g/mol) 36.46
MDL Number MFCD00011324 MFCD00792839
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
Synonym hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
PubChem CID 313
ChEBI CHEBI:17883
IUPAC Name hydrogen chloride
SMILES Cl

Specifications

UN Number UN2924
Concentration 1M in diethyl ether
Density 0.731
Solubility Information Fully misicible in water.
Formula Weight 36.46
Refractive Index 1.3535
Quantity 100 mL
Flash Point −40°C (−40°F)
Sensitivity Air and light sensitive
Chemical Name or Material Hydrochloric acid
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydrogen chloride, 1M in diethyl ether
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 1 A
  • Specific target organ toxicity Category 3
Hazard Statement
  • H224-Extremely flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H336-May cause drowsiness or dizziness.
  • EUH019-May form explosive peroxides.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Ethyl ether, Hydrogen chloride

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.