missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen chloride, 1M soln. in ethyl acetate, Thermo Scientific Chemicals

Catalog Number 15414977
Click to view available options
Quantity:
100 mL
500 mL
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
Cl
7647-01-0
ClH
36.46
MFCD00011324 MFCD00792839
VEXZGXHMUGYJMC-UHFFFAOYSA-N
hydrochloric acid, hydrogen chloride, muriatic acid, chlorohydric acid, acide chlorhydrique, chlorwasserstoff, spirits of salt, hydrogen chloride hcl, anhydrous hydrochloric acid, chloorwaterstof
313
CHEBI:17883
Cl

In chemical industry production of inorganic salts, Production of vinyl chloride from acetylene, alkyl chlorides from olefins, dyes, artificial silk, pigment, pharmaceutical hydrochloride, As catalyst and solvent in organic syntheses, Alcohol chlorination reagent, Nitration reaction, Manufacture of phosphoric acid and in the production of ammonium chloride, Neutralization of basic systems, Cleaning agents. Hydrolyzing of starch and proteins in the preparation of various food products. The hydrogen chloride gas is mainly used for EDC and vinyl chloride monomer (VCM) production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Linear Formula HCl
UN Number UN2924
Concentration 1M soln. in ethyl acetate
Density 0.9
Solubility Information Reacts with water.
IUPAC Name hydrogen chloride
Formula Weight 36.46
Quantity 100 mL
Chemical Name or Material Hydrochloric acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'