missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen chloride, technical, 4 to 6N solution in 2-propanol

Product Code. 15225063 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
This item is not returnable. View return policy

Product Code. 15225063

Brand: Thermo Scientific Acros 450720025

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7647-01-0 | ClH | 36.46 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Hydrochloric acid
CAS 7647-01-0, 67-63-0
Density 0.9090g/mL
Flash Point 11°C
Specific Gravity 0.909
Molecular Formula ClH
Linear Formula HCl
Concentration 4 to 6 N
MDL Number MFCD00011324 MFCD00792839
Solubility Soluble in water, methanol, ethanol and 2-propanol
InChI Key VEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILES Cl
IUPAC Name hydrogen chloride
Molecular Weight (g/mol) 36.46
PubChem CID 313
Quantity 2.5 L
ChEBI CHEBI:17883
Formula Weight 36.45
Grade Technical
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.