missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide 12% (w/v) (40 Volumes), Pure, Fisher Chemical

Product Code. 10776481 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
2.5 L
Unit Size:
2.5 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10776481 2.5 L 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10776481 Supplier Fisher Chemical Supplier No. H/1650/17
 Restricted Item
Estimated Shipment: 19-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

Color Colorless
CAS 7722-84-1
Boiling Point 100°C
Melting Point 0°C
Molecular Formula H2O2
MDL Number 11333
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
SMILES OO
IUPAC Name hydrogen peroxide
Molecular Weight (g/mol) 34.014
PubChem CID 784
ChEBI CHEBI:16240
Packaging HDPE plastic bottle
Quantity 2.5 L
Physical Form Liquid
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • Hydrogen peroxide, 12% w/v
Signal Word
  • Danger
Hazard Category
  • Oxidising liquid Category 3
  • Serious eye damage/eye irritation Category 1
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H318-Causes serious eye damage.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contain: Hydrogen peroxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.