missing translation for 'onlineSavingsMsg'
Learn More

hydrogen peroxide, 3 wt.% solution in water, stabilized

Product Code. 10517824 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
25 mL
100 mL
1 L
Packaging:
Plastic bottle
Unit Size:
1 L
100mL
25mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10517824 100 mL Plastic bottle 100mL
10081254 25 mL Plastic bottle 25mL
10061494 1 L Plastic bottle 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10517824 Supplier Thermo Scientific Acros Supplier No. 426001000
From Chemicals to Chromatography
Use promo code 27991
View all promotions
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 7722-84-1 | H2O2 | 34.014 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1 , 7732-18-5
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Boiling Point 100.0°C
Linear Formula HOOH
Density 1.0100g/mL
Merck Index 15, 4835
Solubility Information Solubility in water: soluble.
Formula Weight 34
Packaging Plastic bottle
Specific Gravity 1.01
Quantity 100 mL
Chemical Name or Material Hydrogen peroxide, 3 wt.% solution in water, stabilized

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.