missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen Peroxide 30-32% (w/w) (100 Volumes), Certified AR for Analysis, Fisher Chemical™

Product Code. 10284000 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
25 L
Packaging:
HDPE plastic bottle
Plastic container
Unit Size:
1 L
2.5 L
25 L
Product Code. Quantity Packaging unitSize
10284000 2.5 L HDPE plastic bottle 2.5 L
10736291 1 L HDPE plastic bottle 1 L
11488333 25 L Plastic container 25 L
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Product Code. 10284000

Brand: Fisher Chemical H/1800/17

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

H2O2, CAS Number-7722-84-1, albone, hioxyl, hydrogen dioxide, inhibine, interox, kastone, oxydol, perhydrol, peroxaan, superoxol, 1L, 30-32%w/w, 108 deg.C, CHEBI:16240, Colorless, MHAJPDPJQMAIIY-UHFFFAOYSA-N, hydrogen peroxide, -33 deg.C, 11333, 34.014, HDPE plastic bottle, 3.3, Liquid, 784, OO, 1.1

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number 11333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

Color Colorless
Boiling Point 108°C
Melting Point -33°C
Assay Percent Range 30-32%w/w
Packaging HDPE plastic bottle
Vapor Density 1.1
pH 3.3
Quantity 2.5 L
Physical Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.