missing translation for 'onlineSavingsMsg'
Learn More

Hydrogen PerOxide Solution, 3 wt% in H2O, Solstice

Product Code. 15613820
Change view
Click to view available options
Quantity:
25 mL
500 mL
4 L
Unit Size:
25mL
4 L
500mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15613820 500 mL 500mL
15658360 25 mL 25mL
15668360 4 L 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15613820 Supplier Honeywell-Fluka Supplier No. 323381500ML
  • ONLINE EXCLUSIVE PRICE
    111.00 €
    104.58 € / 500mL
    Save 6.42 €
    5% Off
 Restricted Item
In Stock
Add to basket
Edge

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.

Contains ∼200 ppm acetanilide as stabilizer, 3 wt. % in H2O

  • Leading the Industry For Over 65 Years
  • Solstice now delivers premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7722-84-1
Molecular Formula H2O2
Molecular Weight (g/mol) 34.014
MDL Number MFCD00011333
InChI Key MHAJPDPJQMAIIY-UHFFFAOYSA-N
Synonym oxydol, perhydrol, superoxol, interox, hydrogen dioxide, inhibine, peroxaan, albone, hioxyl, kastone
PubChem CID 784
ChEBI CHEBI:16240
IUPAC Name hydrogen peroxide
SMILES OO

Specifications

UN Number NONH for all modes of transport
Beilstein 3587191
Density 1g/mL at 25°C
Formula Weight 34.01g/mol
Vapor Pressure 23.3mmHg (30°C)
Vapor Density 1.1 (vs air)
Quantity 500 mL
Chemical Name or Material Hydrogen Peroxide Solution
Product Identifier
  • Hydrogen peroxide solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.