missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydrogen Peroxide Solution, ACS Reagent, Honeywell™
65.67 EUR valid until 2024-12-31
Use promo code "21986" to get your promotional price.
Use promo code "21986" to get your promotional price.
Click to view available options
Quantity:
100 mL
500 mL
4 L
Description
- We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
Specifications
Color | Undesignated |
UN Number | 2014 |
Beilstein | 3587191 |
Concentration | 30wt. % in H2O |
Formula Weight | 34.01g/mol |
Grade | ACS Reagent |
Vapor Pressure | 23.3mmHg (30°C) |
Vapor Density | 1 (vs air) |
Quantity | 500 mL |
Chemical Name or Material | Hydrogen Peroxide Solution, Contains inhibitor |
Safety and Handling
Product Identifier
- Hydrogen peroxide solution
- Danger
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 1
- H302-Harmful if swallowed.
- H318-Causes serious eye damage.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- MIXTURE LIST-Contains : hydrogen peroxide solution
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'