missing translation for 'onlineSavingsMsg'
Learn More

R&D Systems™ Human VTA1 ELISA Kit (Colorimetric)

Product Code. 18610971 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Kit
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18610971 1 Kit 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18610971 Supplier R&D Systems™ Supplier No. NBP260609
 Restricted Item
Estimated Shipment: 01-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Novus product, part of R&D Systems. Kit (Colorimetric)

TRUSTED_SUSTAINABILITY

Specifications

Assay Range 0.188 to 12 ng/mL (example only, lot dependent)
Assay Sensitivity 0.25 ng/mL (example only; lot dependent)
Product Line Novus
Product Type ELISA Kit (Colorimetric)
Sample Type Serum, Plasma, Cell Culture
For Use With (Application) ELISA, Sandwich ELISA
Gene ID (Entrez) 51534
Gene Symbol VTA1
Quantity 1 Kit
Research Discipline Virology Bacteria and Parasites
Storage Requirements Storage is content dependent.
Synonym C6orf55DRG1, Dopamine-responsive gene 1 protein, DRG-1, FLJ27228, homolog of mouse SKD1-binding protein 1, LIP5HSPC228, LYST-interacting protein 5, My012, SBP1chromosome 6 open reading frame 55, SKD1-binding protein 1, vacuolar protein sorting-associated protein VTA1 homolog, Vps20-associated 1 homolog (S. cerevisiae)
Target Human
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.