missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroquinone, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Hydroquinone |
| CAS | 123-31-9 |
| Analyte or Component Names | Hydroquinone |
| Shipping Condition | +20°C |
| Molecular Formula | C6 H6 O2 |
| Certifications/Compliance | ISO 17034 |
| Synonym | Calcium Dobesilate Monohydrate Imp. A (EP), Dobesilate Imp. A (EP), Hydroquinone, 1,4-Benzoquinol, 1,4-Dihydroxybenzene, 1,4-Phenylenediol, 1,4-p-Benzenediol, 4-Hydroxyphenol, Aida, Arctuvin, BQ(H), Benzohydroquinone, Benzoquinol, Black & White Bleaching Cream, Diak 5, Dihydroquinone, Eldopacque, Eldopaque, Eldopaque Forte, Eldoquin, Eldoquin Forte, HE 5, Hydroquinol, NSC 9247, Phiaquin, Quinol, Solaquin Forte, Solution Q, Tecquinol, Tenox HQ, p-Benzenediol, p-Dihydroquinone, p-Dihydroxybenzene, p-Dioxybenzene, p-Hydroquinone, p-Hydroxyphenol, p-Phenylenediol, p-Quin, Calcium Dobesilate Monohydrate Impurity A, Etamsylate Impurity A, Calcium Dobesilate Impurity A |
| InChI Formula | InChI=1S/C6H6O2/c7-5-1-2-6(8)4-3-5/h1-4,7-8H |
| SMILES | Oc1ccc(O)cc1 |
| IUPAC Name | benzene-1,4-diol |
| Show More |
Produkttitel
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.
Ser du en mulighed for forbedring?