missing translation for 'onlineSavingsMsg'
Learn More

Hydroquinonesulfonic acid potassium salt, 98+%

Product Code. p-7044831
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
This item is not returnable. View return policy

Product Code. 11328145

Brand: Thermo Scientific Alfa Aesar L08406.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 21799-87-1 | C6H5KO5S | 228.259 g/mol

It is employed in the synthesis and properties of sulfonated polyarylene ether nitrile copolymers for PEM with high thermal stability.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is employed in the synthesis and properties of sulfonated polyarylene ether nitrile copolymers for PEM with high thermal stability.

Solubility
Soluble in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 21799-87-1
Molecular Formula C6H5KO5S
Molecular Weight (g/mol) 228.259
MDL Number MFCD00007475
InChI Key VKDSBABHIXQFKH-UHFFFAOYSA-M
Synonym potassium 2,5-dihydroxybenzenesulfonate, hydroquinonesulfonic acid potassium salt, potassium2,5-dihydroxybenzenesulfonate, unii-249e3f00ep, benzenesulfonic acid, 2,5-dihydroxy-, monopotassium salt, 2,5-dihydroxybenzenesulfonic acid potassium salt, potassium hydroquinonesulfonate, hydroquinone sulfonic acid potassium salt, hydroquinone sulfonic acid, potassium salt, benzenesulfonic acid, 2,5-dihydroxy-, potassium salt 1:1
PubChem CID 23672329
IUPAC Name potassium;2,5-dihydroxybenzenesulfonate
SMILES C1=CC(=C(C=C1O)S(=O)(=O)[O-])O.[K+]

Specifications

Melting Point ∽251°C (decomposition)
Quantity 100 g
Beilstein 5187027
Solubility Information Soluble in water.
Formula Weight 228.27
Percent Purity ≥98%
Chemical Name or Material Hydroquinonesulfonic acid potassium salt

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.