missing translation for 'onlineSavingsMsg'
Learn More

Hydroxy Tolbutamide, TRC

Product Code. 30011731
Change view
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30011731 25 mg 25mg
17910575 10 mg 10mg
17950915 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30011731 Supplier Toronto Research Chemicals Supplier No. TRCH96985025MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5719-85-7
Molecular Formula C12 H18 N2 O4 S
Molecular Weight (g/mol) 286.35
Synonym 4-Hydroxytolbutamide, N-[(Butylamino)carbonyl]-4-(hydroxymethyl)benzenesulfonamide (ACI), Urea, 1-butyl-3-[(α-hydroxy-p-tolyl)sulfonyl]- (6CI, 7CI, 8CI), 1-Butyl-3-(p-hydroxymethylphenyl)sulfonylurea, 1-Butyl-3-[4-(hydroxymethyl)phenyl]sulfonylurea, Hydroxymethylcarbutamide, Hydroxymethyltolbutamide, Hydroxytolbutamide
IUPAC Name 1-butyl-3-[4-(hydroxymethyl)phenyl]sulfonylurea
SMILES CCCCNC(=O)NS(=O)(=O)c1ccc(CO)cc1

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C12H18N2O4S/c1-2-3-8-13-12(16)14-19(17,18)11-6-4-10(9-15)5-7-11/h4-7,15H,2-3,8-9H2,1H3,(H2,13,14,16)
Formula Weight 286.0987
Percent Purity >95%
Chemical Name or Material 4-Hydroxytolbutamide
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.