missing translation for 'onlineSavingsMsg'
Learn More

[Hydroxy(tosyloxy)iodo]benzene, 97%

Product Code. 10117233 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10117233 5 g Glass bottle 5g
10035293 25 g Glass bottle 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10117233 Supplier Thermo Scientific Acros Supplier No. 389610050
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 27126-76-7 | C13H13IO4S | 392.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 27126-76-7
Molecular Formula C13H13IO4S
Molecular Weight (g/mol) 392.21
MDL Number MFCD00011547
InChI Key LRIUKPUCKCECPT-UHFFFAOYSA-N
Synonym hydroxy tosyloxy iodo benzene, htib, koser's reagent, phenyliodosohydroxy tosylate, koser reagent, unii-2eoa97b14a, hydroxy p-tolylsulfonyl oxy iodo benzene, hydroxy 4-methylbenzenesulfonato-o phenyliodine, iodosobenzene-i-mono-p-toluenesulfonate, hydroxy phenyl 4-methylbenzenesulfonoperoxoyl iodide
PubChem CID 325434
IUPAC Name [hydroxy(phenyl)-$l^{3}-iodanyl] 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OI(C2=CC=CC=C2)O

Specifications

CAS Min % 97.0
CAS Max % 100.0
Melting Point 131.0°C to 137.0°C
Color Yellow
Infrared Spectrum Authentic
Assay Percent Range 97+%
Packaging Glass bottle
Quantity 5 g
Merck Index 15, 5369
Formula Weight 392.21
Percent Purity 97+%
Physical Form Crystalline Powder
Chemical Name or Material [Hydroxy(tosyloxy)iodo]benzene
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.