missing translation for 'onlineSavingsMsg'
Learn More

Hydroxychloroquine sulfate, MedChemExpress

Product Code. 16464587
Encompass_Preferred
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16464587 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16464587 Supplier Medchem Express Supplier No. HYB1370
 Restricted Item
Estimated Shipment: 27-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hydroxychloroquine sulfate (HCQ sulfate) is a synthetic antimalarial agent which can also inhibit Toll-like receptor 7/9 (TLR7/9) signaling. Hydroxychloroquine sulfate is efficiently inhibits SARS-CoV-2 infection in vitro.

Hydroxychloroquine sulfate (HCQ sulfate) is a synthetic antimalarial agent which can also inhibit Toll-like receptor 7/9 (TLR7/9) signaling. Hydroxychloroquine sulfate is efficiently inhibits SARS-CoV-2 infection in vitro.

Specifications

Chemical Name or Material Hydroxychloroquine sulfate
Physical Form Solid
CAS 747-36-4
Color White
Quantity 50 mg
Purity Grade Notes Research
Molecular Formula C18H28ClN3O5S
For Use With (Application) Cancer-programmed cell death
Synonym HCQ sulfate
Solubility Information H2O : 110 mg/mL (253.49 mM; Need ultrasonic and warming) ∣DMF : 1.4 mg/mL (3.23 mM; Need ultrasonic) ∣DMSO : < 1 mg/mL (ultrasonic) (insoluble or slightly soluble)
SMILES CCN(CCO)CCCC(NC1=CC=NC2=CC(Cl)=CC=C12)C.O=S(O)(O)=O
Molecular Weight (g/mol) 433.95
Formula Weight 433.95
Percent Purity 98.68%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.