missing translation for 'onlineSavingsMsg'
Learn More

Hydroxylamine Alcoholic solution EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA

Product Code. 12950434 Shop All Fisher Chemical Products
Click to view available options
Quantity:
1000 mL
Unit Size:
100mL
This item is not returnable. View return policy

Product Code. 12950434

Brand: Fisher Chemical RP 097 100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.

Specifications

Packaging Bottle
Analyte or Component Names Hydroxylamine solution, alcoholic
Purity Grade Notes EU Ph Ref: 1044301
Element Hydroxylamine
Solvent or Matrix Hydrogen Chloride/Hydrogen Fluoride/Nitric Acid
Quantity 1000 mL
Grade Eu. Ph.
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hydroxylamine solution,alcoholic. European Pharmacopoeia Ref: 1044301 (RP 097)
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
  • Skin sensitiser Category 1
  • Carcinogenicity Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H317-May cause an allergic skin reaction.
  • H351-Suspected of causing cancer.
Precautionary Statement
  • P202-Do not handle until all safety precautions have been read and understood.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P308+P313-IF exposed or concerned: Get medical advice/attention.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P370+P378-In case of fire: Use to extinguish.
  • P403+P235-Store in a well-ventilated place. Keep cool.
  • P501-Dispose of contents/container to .
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.