missing translation for 'onlineSavingsMsg'
Learn More

Hydroxyurea, MedChemExpress

Product Code. 16490868
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490868 5 g 5g
16482129 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16490868 Supplier Medchem Express Supplier No. HYB0313
 Restricted Item
Estimated Shipment: 06-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hydroxyurea is a cell apoptosis inducer that inhibit DNA synthesis through inhibition of ribonucleotide reductase. Hydroxyurea shows anti-orthopoxvirus activity.

Hydroxyurea is a cell apoptosis inducer that inhibit DNA synthesis through inhibition of ribonucleotide reductase. Hydroxyurea shows anti-orthopoxvirus activity.

Specifications

Chemical Name or Material Hydroxyurea
Physical Form Solid
CAS 127-07-1
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula CH4N2O2
For Use With (Application) Cancer-programmed cell death
Synonym Hydroxycarbamide
Solubility Information DMSO : 100 mg/mL (1314.92 mM; Need ultrasonic) ∣H2O : 50 mg/mL (657.46 mM; Need ultrasonic)
SMILES O=C(N)NO
Molecular Weight (g/mol) 76.05
Formula Weight 76.05
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.