missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Hydroxyzine dihydrochloride, MedChemExpress
Description
Hydroxyzine dihydrochloride, a benzodiazepine antihistamine agent, acts as a orally active histamine H1-receptor and serotonin antagonist. Hydroxyzine dihydrochloride has anxiolytic effect and can be used forthe research of generalised anxiety disorder.
Specifications
Specifications
| Chemical Name or Material | Hydroxyzine dihydrochloride |
| Physical Form | Solid |
| CAS | 2192-20-3 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C21H29Cl3N2O2 |
| For Use With (Application) | COVID-19-immunoregulation |
| Solubility Information | H2O : ≥ 150 mg/mL (334.95 mM) |
| SMILES | ClC1=CC=C(C=C1)C(C2=CC=CC=C2)N3CCN(CCOCCO)CC3.Cl.Cl |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?