Learn More
Fisher BioReagents™ Hygromycin B Aminoglycoside Antibiotic
Brand:  Fisher BioReagents™ 12891600
Code : 32
Additional Details : CAS Number : 31282-04-9 Weight : 0.10000kg Transport : UN number : 2810 Chem class : 6.1 Pack group : II
Product Code. 12891600
  • 837.40 € / 0.10 L
Estimated Shipment: 05-07-2024
to see stock.
Description
Description
Useful as selective agent for the APH4 gene in plant tissue supplied as 1M units in solution, 1 vial: 5 x 106 Fisher BioReagents
Specifications
Specifications
1M units