missing translation for 'onlineSavingsMsg'
Learn More

Hygromycin B

Product Code. 15403419
Change view
Click to view available options
Quantity:
100 mg
250 mg
1 g
Unit Size:
100mg
1g
250mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15403419 100 mg 100mg
15413419 250 mg 250mg
15493409 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15403419 Supplier Thermo Scientific Alfa Aesar Supplier No. J60681.MC
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

Hygromycin B, CAS # 31282-04-9, is an aminoglycoside compound showing antibiotic activity that kills bacteria through the inhibition of protein synthesis. It also presents anthelmintic activity. | CAS: 31282-04-9 | C20H37N3O13 | 527.524 g/mol

An aminoglycoside antibiotic that kills bacteria by inhibiting protein synthesisHygromycin B is used for the selection and maintenance of prokaryotic and eukaryotic cells that contains the hygromycin resistance gene. This gene is frequently used as a selectable marker in research on plants. It stabilizes the transfer ribonucleic acid (tRNA)-ribosomal acceptor site, thereby inhibiting translocation. In addition to this, it serves as an aminoglycosidic antibiotic which inhibits protein synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Hygromycin is an aminoglycosidic compound that shows antibiotic activity. It is produced by Streptomyces hygroscopicus
  • Hygromycin acts against both prokaryotic and eukaryotic cells, having the ability to inhibit protein synthesis. It also stabilizes the transfer ribonucleic acid (tRNA)-ribosomal acceptor site, thereby inhibiting translocation. This compound prevents ribosome translocation during translation elongation by binding the 30S rRNA proximal to the ribosomal E, P, and A sites

Application

  • In clinical and plant research, this compound can be used as a marker to select and maintain prokaryotic and eukaryotic cells containing the hygromycin resistance gene
  • Hygromycin also presents an anthelmintic activity against nematode parasites of chickens
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 31282-04-9
Molecular Formula C20H37N3O13
Molecular Weight (g/mol) 527.524
MDL Number MFCD06795479
InChI Key GRRNUXAQVGOGFE-BBMONYMYSA-N
PubChem CID 134129613
IUPAC Name (3'S,3aS,4S,4'S,5'R,6R,6'R,7R,7aS)-4-[(1R,2S,3R,5S,6S)-3-amino-2,6-dihydroxy-5-(methylamino)cyclohexyl]oxy-6'-[(1S)-1-amino-2-hydroxyethyl]-6-(hydroxymethyl)spiro[4,6,7,7a-tetrahydro-3aH-[1,3]dioxolo[4,5-c]pyran-2,2'-oxane]-3',4',5',7-tetrol
SMILES CNC1CC(C(C(C1O)OC2C3C(C(C(O2)CO)O)OC4(O3)C(C(C(C(O4)C(CO)N)O)O)O)O)N

Specifications

Type Hygromycin B
Physical Form Lyophilized Powder
Color Brown
Quantity 100 mg
UN Number UN3462
Merck Index 14,4852
Solubility Information Miscible with water,methanol,buffer solution and ethanol; Immiscible with less polar solvents.
Formula Weight 527.53
Chemical Name or Material Hygromycin B
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Hygromycin B
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 1
  • Acute toxicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Respiratory sensitiser Category 1
  • Skin sensitiser Category 1
Hazard Statement
  • H300-Fatal if swallowed.
  • H310-Fatal in contact with skin.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H330-Fatal if inhaled.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P330-Rinse mouth.
Supplemental information
  • MIXTURE LIST-Contain: Hygromycin B

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.