missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners, 300L Liner - Diameter: 24in. (61cm)

Product Code. 30040091 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
Each
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30040091 Each Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30040091 Supplier Thermo Scientific™ Supplier No. SH30647.06
  • ONLINE EXCLUSIVE PRICE
    1840.00 €
    1558.03 € / Pack of 10
    Save 281.97 €
    15% Off
 Restricted Item
In Stock
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo Scientific™ HyPerforma™ 50 to 500L Tank Liners are available with or without bottom drain and are provided sterile for handling fluids in bioproduction applications.

Tank Liner Key Features:

  • Available in sizes that fit into industry standard cylindrical tanks and in Thermo Scientific HyPerforma Drums (SV3011X, SV50076, and SV50517 series)
  • Gusset design allows for a close fit to the tank
  • Chamber constructed of Thermo Scientific CX3-9 film, which provides the same contact surface as other Thermo Scientific BPCs
  • Supplied gamma irradiated
  • Customization options are available by contacting one of our Thermo Scientific Representatives today

Standard Tank Liners for your liquid applications available in-stock in the following configurations:

SH30647.01 to .03:

  • 50 to 200L Tank Liner
  • Support Container Needed: Design to fit into HyPerforma Flat Bottom PE Drums (SV50076 series)
  • Quantity: Each = 1 Liner

SH30647.04 to .07:

  • 50 to 500L Tank Liner
  • Support Container Needed: Designed to fit into Industry Standard Cylindrical Tanks (SV3011X series)
  • Quantity: Each = 1 Liner

SH30646 Series:

  • 50 to 200L Tank Liner with One Bottom Drain
  • Bottom Lineset 1: 72in. (182.9cm) of tubing leading to a Quick Disconnect (MPX series) Insert (male) connector
  • Support Container Needed: Designed to fit into HyPerforma Conical Bottom PE Drums (SV50517 series)

Ordering Information:

Supplied double polybagged and irradiated with Certificate of Analysis.

Recommended for:

  • Hydrations of powdered media and buffers
  • Pooling of non-sterile solutions and fluids
  • Non-aseptic collection of waste liquids from separation and purification systems

To order, use the part numbers listed in the above table.

TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 300 L
Outer Packaging Packages are placed in strong cardboard cartons with product and lot identification.
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area.
Quantity Each
For Use With (Equipment) SV30114
Sterilization Method Irradiated
Product Line HyPerforma
Film Description CX3-9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.