missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyPerforma™ Single-Use Mixer DS 300, Unit with Drum Positioner and Adjustable Motor Head

Product Code. 30214829 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
300 L
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30214829 300 L Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30214829 Supplier Thermo Scientific™ Supplier No. SUMDS0300.9003
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

Les retours ne sont pas autorisés pour ce produit. Afficher la politique du retour.
Règlementaire : Cet article est soumis à éco-contribution.

The Thermo Scientific™ HyPerforma™ DS 300 Single-Use Mixer (S.U.M.) is built upon the proven legacy design of the S.U.M; but optimized for lower-cost, high throughput applications. The smaller footprint and lighter weight of the HyPerforma S.U.M. DS 300 provide mobility and flexibility in mixing applications not currently available in the market.

  • The docking station consists of a stainless steel base with locking casters, an adjustable handle, a tethered hand-held control device, an electronic vertical lift mechanism with integrated height indicator, motor mount and motor, three sizes of drive shafts, two adjustment tools - a spanner and a torque wrench. An adjustable-angle motor head and drum positioned are available as add-on options.
  • Supporting hardware optimized for mixing, storage, transport and discharge in a closed system to help minimize the risk of cross-contamination.
  • Top-drain support containers are available in 50, 100, 200 and 300 L sizes; with bottom-drain drums available in 50, 100 and 200 L sizes.
  • Standard BPCs available for liquid/liquid and powder/liquid mixing.
  • Optional dollies are available for all drum sizes.

Applications of the Thermo Scientific HyPerforma DS 300 S.U.M. include:

  • Hydration of powdered media and buffers.
  • Sterile mixing of buffers, cell culture media, or other process fluids.
  • Pooling of sterile liquid fractions.

Accessory: Reusable Bearing Hub, Cat # SV5017777

TRUSTED_SUSTAINABILITY

Spécification

Capacity (Metric) 300 L
Material Stainless Steel (304)
Mixing Rate Range 30-350 RPM
Quantity 300 L
Power Requirements 108 - 240 VAC 50- 60 Hz; 15 amps
Weight 180 kg (398 Ib.)
Product Line HyPerforma
Nom du produit
Sélectionnez un problème

En cliquant sur Soumettre, vous reconnaissez que vous pouvez être contacté par Fisher Scientific au sujet des informations que vous avez fournies dans ce formulaire. Nous ne partagerons pas vos informations à d'autres fins. Toutes les informations de contact fournies seront également conservées conformément à notre politique de confidentialité. Politique de confidentialité.