missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyPerforma Tank Liners, CX3-9 Liner

Product Code. 17810647 Shop All Thermo Scientific Products
Change view
Click to view available options
Capacity (Metric):
19 L
28 L
38 L
378 L
757 L
Unit Size:
Pack of 10
Pack of 5
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) unitSize
17810647 378 L Pack of 10
17820567 757 L Pack of 10
17840567 19 L Pack of 10
17830567 28 L Pack of 10
17870567 38 L Pack of 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 17810647 Supplier Thermo Scientific™ Supplier No. SH30647.10
 Restricted Item
Estimated Shipment: 22-10-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Tank liner system with top train is composed of CX3-9 single web, multi-layer plastic film, with dimensions optimized for drums and commonly used industry standard cylindrical tanks. They feature an open top for maximum recovery, with drum dolly available to provide mobility.

  • Minimizes bioburden due to construction in an ISO 7 cleanroom under sterile cGMP conditions.
  • Eliminates post-use cleaning steps required with re-useable containers and reduces cross-contamination risks.

Applications:

  • Removes the need for tank cleaning and helps reduce cycle times.
  • Facilitates hydration of powdered media and buffers, and pooling of non-sterile solutions and fluids.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (Metric) 378 L
Capacity (English) 100 gal.
Outer Packaging Cardboard Cartons
Protective Inner Packaging Packed and sealed in two independent outer layers in the Class 10,000/ISO Class 7 area
Sterilization Method Irradiated
Product Line HyPerforma
Film Description CX3-9
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.