missing translation for 'onlineSavingsMsg'
Learn More

Hypericin, TRC

Product Code. 17983695
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
10 mg
Unit Size:
10mg
1mg
2.50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
17983695 2.5 mg 2.50mg
17922045 10 mg 10mg
17904775 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17983695 Supplier Toronto Research Chemicals Supplier No. TRCH9981902.5MG
 Restricted Item
Estimated Shipment: 29-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 548-04-9
Molecular Formula C30 H16 O8
Molecular Weight (g/mol) 504.44
Synonym Phenanthro[1,10,9,8-opqra]perylene-7,14-dione, 1,3,4,6,8,13-hexahydroxy-10,11-dimethyl-, stereoisomer (6CI, 7CI, 8CI, 9CI, ACI), stereoisomer of 1,3,4,6,8,13-Hexahydroxy-10,11-dimethylphenanthro[1,10,9,8-opqra]perylene-7,14-dione (ACI), 1,3,4,6,8,13-Hexahydroxy-10,11-dimethylphenanthro[1,10,9,8-opqra]perylene-7,14-dione P-conformer, Cyclo-Werol, Cyclosan, HyBryte, Hyperflav, Hypericin, Hypericum red, NSC 407313, SGX 301
SMILES O=C1C2=C(O)C=C(O)C=3C=4C(O)=CC(O)=C5C(=O)C6=C(O)C=C(C7=C6C(C54)=C(C23)C8=C1C(O)=CC(=C87)C)C

Specifications

Quantity 2.5 mg
InChI Formula InChI=1S/C30H16O8/c1-7-3-9(31)19-23-15(7)16-8(2)4-10(32)20-24(16)28-26-18(12(34)6-14(36)22(26)30(20)38)17-11(33)5-13(35)21(29(19)37)25(17)27(23)28/h3-6,31-36H,1-2H3
Formula Weight 504.0845
Percent Purity >95%
Chemical Name or Material Hypericin
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.