missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypersil™ BDS C8 HPLC Columns

Product Code. 10672854 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
50 mm
100 mm
125 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
3 mm
4 mm
4.6 mm
Particle Size:
2.4 μm
3 μm
5 μm
Unit Size:
Each
Pack of 4
30 product options available for selection
Product selection table with 30 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Particle Size unitSize
10672854 50 mm 2.1 mm 5 μm Each
10448184 50 mm 3 mm 5 μm Each
10394173 50 mm 4.6 mm 5 μm Each
10110593 100 mm 2.1 mm 3 μm Each
10582255 100 mm 2.1 mm 5 μm Each
10354223 100 mm 4.6 mm 5 μm Each
10680136 100 mm 4 mm 3 μm Each
10763016 150 mm 4 mm 5 μm Each
11583061 125 mm 4 mm 5 μm Each
10786153 150 mm 4.6 mm 5 μm Each
10476664 250 mm 4 mm 5 μm Each
10364653 250 mm 4.6 mm 5 μm Each
10354653 10 mm 4 mm 5 μm Pack of 4
10662854 10 mm 3 mm 5 μm Pack of 4
10099132 150 mm 4.6 mm 3 μm Each
10332675 150 mm 3 mm 3 μm Each
10533215 100 mm 4.6 mm 3 μm Each
10078942 50 mm 4.6 mm 3 μm Each
10600364 50 mm 2.1 mm 3 μm Each
10512065 10 mm 4 mm 3 μm Pack of 4
10747883 10 mm 3 mm 3 μm Pack of 4
10610764 10 mm 2.1 mm 3 μm Pack of 4
10584285 150 mm 4.6 mm 2.4 μm Each
10457514 150 mm 2.1 mm 2.4 μm Each
10137604 100 mm 4.6 mm 2.4 μm Each
10239914 100 mm 2.1 mm 2.4 μm Each
10343105 50 mm 2.1 mm 2.4 μm Each
11969108 10 mm 4 mm 2.4 μm Pack of 4
11919108 10 mm 2.1 mm 2.4 μm Pack of 4
10100883 10 mm 2.1 mm 5 μm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 30 options available.
30 options
This item is not returnable. View return policy
Product Code. 10672854 Supplier Thermo Scientific™ Supplier No. 28205052130
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

Use as a robust, general-purpose column for QC/QA applications. They are high-quality, base-deactivated, fully endcapped, have similar C18 selectivity but slightly less retentive.

For a high-quality, base-deactivated, fully endcapped phase with similar selectivity to C18 but slightly less retention, use Thermo ScientificTM HypersilTM BDS C8 HPLC Columns. These economical columns feature reduced silanol activity and provide exceptional reliability for QA/QC labs. Built on the renowned Hypersil silica backbone, the base-deactivated silica is one of the most popular packing materials available.

For more challenging separations that require improved chromatographic performance, use Thermo ScientificTM Hypersil GOLDTM C8 columns as an alternative.

  • Excellent choice for acids, bases and neutral compounds
  • Less retentive than C18 for faster analyses
  • Robust columns with long column lifetimes
  • Very reliable for routine assays

Ordering Information:

Other column dimensions are available. Please call Customer Service for more information.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 2.1 mm
Length (Metric) 50 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 130 Å
Surface Area 170 m²/g
Temperature 60°C
USP Type L7
Stationary Phase C8
Product Line Hypersil BDS
Packing Material Spherical, Fully Porous, Base-Deactivated Silica
Carbon Load 7%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.