missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypersil GOLD™ C4 HPLC Columns

Product Code. 10688435 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
10 mm
30 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
1 mm
2.1 mm
3 mm
4 mm
4.6 mm
Particle Size:
1.9 μm
3 μm
5 μm
Unit Size:
Each
Pack of 4
28 product options available for selection
Product selection table with 28 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Particle Size unitSize
10688435 150 mm 4 mm 5 μm Each
10609585 50 mm 2.1 mm 5 μm Each
10699385 50 mm 4.6 mm 5 μm Each
10382865 100 mm 2.1 mm 5 μm Each
10322355 100 mm 3 mm 5 μm Each
10699755 100 mm 4.6 mm 5 μm Each
10609765 150 mm 2.1 mm 5 μm Each
10486854 150 mm 4.6 mm 5 μm Each
10067684 250 mm 2.1 mm 5 μm Each
10312635 250 mm 4.6 mm 5 μm Each
10127614 50 mm 2.1 mm 1.9 μm Each
10691396 100 mm 2.1 mm 1.9 μm Each
10391585 150 mm 2.1 mm 1.9 μm Each
10147884 10 mm 1 mm 3 μm Pack of 4
10447534 10 mm 2.1 mm 3 μm Pack of 4
10157884 10 mm 3 mm 3 μm Pack of 4
10610626 10 mm 4 mm 3 μm Pack of 4
10332835 30 mm 2.1 mm 3 μm Each
10159534 50 mm 2.1 mm 3 μm Each
10037254 100 mm 2.1 mm 3 μm Each
10027504 100 mm 3 mm 3 μm Each
10732826 100 mm 4.6 mm 3 μm Each
10199474 150 mm 1 mm 3 μm Each
10599735 150 mm 2.1 mm 3 μm Each
10312355 150 mm 3 mm 3 μm Each
10261325 150 mm 4.6 mm 3 μm Each
10076694 10 mm 2.1 mm 5 μm Pack of 4
10670046 10 mm 4 mm 5 μm Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 28 options available.
28 options
This item is not returnable. View return policy
Product Code. 10688435 Supplier Thermo Scientific™ Supplier No. 25505154030

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Featuring a lower hydrophobicity than C18 or C8 columns, Thermo Scientific™ Hypersil GOLD™ C4 columns provide outstanding sensitivity and high efficiency.

For very hydrophobic analytes, achieve faster separations with excellent peak shapes using Thermo Scientific™ Hypersil GOLD™ C4 HPLC columns. Featuring a lower hydrophobicity than C18 or C8 columns, Hypersil GOLD C4 columns provide outstanding sensitivity and high efficiency.

Similar Selectivity, Less Retention Than C18 or C8 Columns

  • Successful retention and separation of very hydrophobic analytes with this short alkyl chain length, low hydrophobicity column
  • Ideal for analysis of hydrophobic polypeptides and proteins
  • Superior Performance
  • Highly symmetrical peak shapes 
  • 1.9μm particle size columns improve speed and efficiency

About the Hypersil GOLD Family of HPLC Columns

Meet the challenges of modern chromatography with state-of-the-art columns that provide outstanding performance and consistent separations. Twelve different chemistries are available so you can optimize separations and maximize productivity. Get outstanding peak shapes in reversed phase, ion exchange, HILIC or normal phase chromatography.

  • Excellent peak symmetry
  • Narrow peaks for outstanding efficiency
  • Enhanced sensitivity and improved resolution
  • Exceptional reproducibility for reliable chromatography, column after column
  • Solutions for high-throughput screening
  • 1.9, 3 and 5μm particle sizes for all your separation needs
TRUSTED_SUSTAINABILITY

Specifications

Diameter (Metric) 4 mm
Flow Rate 0.8 mL/min
Length (Metric) 150 mm
Max. Pressure 5800 psi (400 bar)
Particle Size 5 μm
pH 2 to 8
Pore Size 175 Å
Surface Area 220 m²/g
Temperature 60°C
USP Type L26
Stationary Phase C4
Packing Material Silica
Carbon Load 5%
Column Type Reversed Phase
Endcapped Yes
Phase Reversed Phase
Show More Show Less

Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.