missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Hypersil GOLD™ Peptide HPLC Column

Product Code. 17916092 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
50 mm
100 mm
150 mm
Max. Pressure:
14503 psi (1000 bar)
21756 psi (1500 bar)
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Max. Pressure Unit Size
17916092 150 mm 21756 psi (1500 bar) Each
17906092 50 mm 14503 psi (1000 bar) Each
17926092 100 mm 21756 psi (1500 bar) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17916092 Supplier Thermo Scientific™ Supplier No. 26002152130
Big Savings for Better Separation
Use promo code 27992
View all promotions
 Restricted Item
Estimated Shipment: 20-10-2026
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Hypersil GOLD™ Peptide performance is assured by our dual quality confirmation, which is performed on both neutrals compounds as well as on a Cytochrome C digest.

Resolve peptide challenges with one column - NEW Hypersil GOLD Peptide.

Achieve consistent retention of all peptides, including hydrophilic ones, and resolve deamidated species. Designed for stability and lot-to-lot consistency, this column meets the needs of biopharmaceutical product development and manufacturing control.

Hypersil GOLD Peptide column performance is assured through dual quality confirmation, including a peptide-based QC test, ensures confidence in ultrapure silica, ligand bonding, and column packing for unparalleled sequence coverage and higher peak capacity compared to other peptide columns.

Consistent, Reliable Results

Enjoy excellent reproducibility with minimal RSD%, ensuring consistency from column-to-column and lot-to-lot.

Optimized Peptide Retention - Retain More, Discover More

Our column uniquely holds onto hydrophilic peptides, ensuring no critical data is missed.

Exceptional Peak Capacity - Separate with Superiority

Achieve higher peak capacity for detailed and clear separations, ensuring every component is distinctly resolved.

Advanced PTM Analysis - Decipher Complex Modifications

Efficiently separate key post-translational modifications like deamidation, crucial for in-depth protein analysis. Excellent peak shape for peptides using either FA- or TFA-containing mobile phases.

Unparalleled Sequence Coverage - See the Full Picture

High sequence coverage provides a comprehensive analysis, revealing the true complexity of your samples.

TRUSTED_SUSTAINABILITY

Specifications

Column Format Analytical Column
Diameter (Metric) 2.1 mm
For Use With UHPLC
Length (Metric) 150 mm
Max. Pressure 21756 psi (1500 bar)
Particle Shape Fully Porous
Particle Size 1.9 μm
pH 2 to 11
Pore Size 175 Å
Quantity Each
Surface Area 220 m²/g
Temperature 60°C
USP Type L1
Stationary Phase C18 Selectivity Peptide
Product Line Hypersil GOLD
Packing Material Ultrapure Silica
Carbon Load 11%
Endcapped Yes
Phase Reversed Phase
For Use With (Application) Peptide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.