missing translation for 'onlineSavingsMsg'
Learn More

Hypromellose, MedChemExpress

Product Code. 16422019
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16422019 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16422019 Supplier Medchem Express Supplier No. HYA0104
 Restricted Item
Estimated Shipment: 21-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Hypromellose is a hydrophilic, non-ionic cellulose ether used to form swellable-soluble matrices.

Hypromellose is a hydrophilic, non-ionic cellulose ether used to form swellable-soluble matrices.

Specifications

Chemical Name or Material Hypromellose
Physical Form Solid
CAS 9004-65-3
Color White
Quantity 1 g
Purity Grade Notes Research
Synonym (Hydroxypropyl)methyl cellulose HPMC Celacol HPM 5000
Solubility Information DMSO : 25 mg/mL (Need ultrasonic) ∣H2O : 10 mg/mL (Need ultrasonic)
SMILES [R]O[C@@H]1[C@H](O[R])[C@@H](O[R])[C@H](O[C@H]2[C@H](O[R])[C@@H](O[R])[C@H](O[R])[C@@H](CO[R])O2)[C@@H](CO[R])O1.CO.CC(O)CO.[n].[x]
Grade Research

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.