missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ HyPURITY™ C18 HPLC Columns, 3μm Particle Size

Product Code. 10761937 Shop All Thermo Scientific Products
Change view
Click to view available options
Length (Metric):
30 mm
50 mm
100 mm
150 mm
250 mm
Diameter (Metric):
2.1 mm
3 mm
4 mm
4.6 mm
Particle Size:
3 μm
5 μm
Unit Size:
Each
27 product options available for selection
Product selection table with 27 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Diameter (Metric) Particle Size Unit Size
10761937 - - - -
10096084 50 mm 2.1 mm 3 μm Each
10648445 50 mm 4.6 mm 3 μm Each
10109194 100 mm 2.1 mm 3 μm Each
10483285 100 mm 3 mm 3 μm Each
10290755 100 mm 4.6 mm 3 μm Each
10648615 150 mm 2.1 mm 3 μm Each
10118954 150 mm 3 mm 3 μm Each
10392965 150 mm 4.6 mm 3 μm Each
10704437 - - - -
10006114 - - - -
10690236 - - - -
10751937 - - - -
10017514 30 mm 2.1 mm 5 μm Each
10477634 50 mm 2.1 mm 5 μm Each
10230615 50 mm 3 mm 5 μm Each
10077554 50 mm 4.6 mm 5 μm Each
10231195 100 mm 2.1 mm 5 μm Each
10270905 100 mm 4 mm 5 μm Each
10408384 100 mm 4.6 mm 5 μm Each
10601116 150 mm 2.1 mm 5 μm Each
10067644 150 mm 3 mm 5 μm Each
10399243 150 mm 4 mm 5 μm Each
10630346 150 mm 4.6 mm 5 μm Each
10518795 250 mm 2.1 mm 5 μm Each
10077454 250 mm 4 mm 5 μm Each
10528795 250 mm 4.6 mm 5 μm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 27 options available.
27 options
This item is not returnable. View return policy
Product Code. 10761937 Supplier Thermo Scientific™ Supplier No. 22105014001
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Thermo ScientificTM HyPURITYTM C18 HPLC Columns are available in limited dimensions. Please contact Customer Service for more details.

  • The 10mm design offers maximum protection with minimal increase in retention
  • Fit Thermo Scientific UNIGUARD™ direct connection and stand alone holders

Features of HyPURITY Solid Phase

  • High quality, highly pure silica
  • Highly characterized, tested and validated C18 columns

Requires:

Thermo Scientific UNIGUARD direct connection or stand alone holder, sold separately.

TRUSTED_SUSTAINABILITY
Product Title
Seleccione un problema

Al hacer clic en Enviar, acepta que Fisher Scientific se ponga en contacto con usted en relación con los comentarios que ha proporcionado en este formulario. No compartiremos su información para ningún otro fin. Toda la información de contacto proporcionada se mantendrá de acuerdo con nuestra Política de Privacidad. Política de privacidad.